Palladium 20% on activated carbon powder standard wetted with ca. 50% water, unreduced

Supplier: Thermo Fisher Scientific

38309.06 38309.14 38309.22
38309.06EA 382 EUR
38309.06 38309.14 38309.22
Palladium 20% on activated carbon powder standard wetted with ca. 50% water, unreduced
Palladium

Physical Form: 900-1100 m²/g Odour: Odourless


Ordering information: Sold on a dry weight basis. Unit weight excludes water weight

Formula: Pd
MW: 106.420 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011167
CAS Number: 7440-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR