Silver nitrate concentrate for dilution to 1L, 0.1 M AgNO3 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Chemicals
Danger

38310-1EA 38310-6X1EA
38310-1EAEA 105 EUR
38310-1EA 38310-6X1EA
Silver nitrate concentrate for dilution to 1L, 0.1 M AgNO3 (0.1N) volumetric solution, Fluka™
Silver nitrate

Concentrate for 1L standard solution

Formula: AgNO₃
Boiling Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3264
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR