Ruthenium 0,5% on carbon, granules

Supplier: Thermo Fisher Scientific

38332.14 38332.22
38332.14EA 263 EUR
38332.14 38332.22
Ruthenium 0,5% on carbon, granules
Ruthenium

CAS No.: 7440-18-8 Physical Form: 4-12 Mesh, 900-1100m2/g Odour: Odourless UN No.: UN1325 MDL No.: MFCD00011207

Formula: Ru
Storage Temperature: Ambient
MDL Number: MFCD00011207
CAS Number: 7440-18-8
EINECS: 231-127-1
UN: 1325
ADR: ,

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR