Salicylic acid 99+%, meetSpec_Ph.Eur.

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 2-Hydroxybenzoic acid, o-Hydroxybenzoic acid

385730010 385730050
385730010.EA 62.8 EUR
385730010. 385730050.
Salicylic acid 99+%, meetSpec_Ph.Eur.
Salicylic acid
Formula: (HO)C₆H₄CO₂H
MW: 138.12 g/mol
Boiling Pt: 211°C (20mmHg)
Flash Pt: 157°C
Storage Temperature: Ambient
MDL Number: MFCD00002439
CAS Number: 69-72-7
EINECS: 200-712-3
Merck Index: 13,08411

Order Now

Specification Test Results

Appearance white to almost white or colorless powder or crystals
Infrared spectrum Conforms
Titration with NaOH 99.0 to 100.5 %
Melting point 158°C to 161°C
Related substances ≤0.1 % (4-Hydroxybenzoic acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR