Methyl ethyl ketone ACS

Supplier: Thermo Fisher Scientific
Danger

389430010 389430025 389430100
389430010.EA 63.4 EUR
389430010. 389430025. ACRO389430100
Methyl ethyl ketone ACS
Methyl ethyl ketone
Formula: C₂H₅COCH₃
Boiling Pt: 80 °C (760 mmHg)
Melting Pt: –87 °C
Density: 0,806 g/cm³
Flash Pt: –7 °C
Storage Temperature: Ambient
MDL Number: MFCD00011648
CAS Number: 78-93-3
EINECS: 201-159-0
UN: 1193
ADR: 3,II
REACH: 01-2119457290-43
Merck Index: 13,06097
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b6656888f6a9a4c0c5ed3053bb2a3dcd8b3af66&vid=17cb98abf677b90f8bac752664d5c0cfd6765591&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
Water ≤0.20 %
GC ≥99.0 %
Color scale ≤15 APHA
Residue after evaporation ≤0.0025 %
Titrable acid ≤0.0005 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR