Cooled incubators, Series 1A
Supplier: LMS
280NP 120 280 80 120NP 280A 80NP 120A 80A 201 201A 201NP
390-0541EA 7190 EUR
390-0541 390-0532 390-0539 390-0542 390-0534 390-0540 390-0544 390-0533 390-0543 390-0536 390-0537 390-0538
Cooled incubators, Series 1A
Incubators Cooled Incubators
LMS Series 1A cooled incubators are temperature controlled cabinets with fan assisted air circulation via a pre-mixing chamber. The units have a white enamelled sheet steel exterior, white pre-coated aluminium interior and polyurethane foam insulation, and feature a door lock and magnetic door gasket. The inner chamber is easy to clean, disinfection procedures are detailed in the user manual.
  • Easy to use controls with digital display of actual temperature on the standard controller or display of both set point and actual temperature on the programmable
More Product Information