VWR® Cellulose Stoppers
Supplier: VWR Collection
391-0151EA 55.8 EUR
391-0151 391-0168 391-0150 391-0152 391-0153 391-0154 391-0161 391-0162 391-0163 391-0164 391-0165 391-0149 391-0167 391-0187 391-0169 391-0173 391-0174 391-0175 391-0176 391-0177 391-0178 391-0179 391-0184 391-0185 391-0186 391-0166 391-0194 391-0191 391-0192 391-0393 391-0412 391-0397 391-0396 391-0392 391-0391 391-0379 391-0377 391-0376 391-0395 391-0381 391-0389 391-0386 391-0382 391-0414 391-0647
VWR® Cellulose Stoppers
Stoppers Cellulose Stoppers
These stoppers are made of cellulose and are used for microbiological samples and tissue cultures in test tubes or Erlenmeyer flasks.
  • Air permeable
  • Autoclavable up to 200 °C
  • Filters bacteria
Ordering information: Available in special or standard shape.
Packaging: Packed in bags and cardboard boxes for easy handling and space saving.
Order Now
SPECIFICATIONS
Environmentally Preferable