Cloning cylinders, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
H378470000 F378470100 F378470200 F378470300
BELAH378470000EA 361 EUR
BELAH378470000 391-1987 391-1988 391-1989
Cloning cylinders, SP Bel-Art
Cylinders Cloning Cylinders
These cloning cylinders facilitate the isolation of individual cell clones, each generated from a single cell, yielding a homogenous population of cells.
  • Cylinders are tapered so bottom edge is wider than the top ensuring a good sealing surface
  • Polystyrene cloning cylinders are supplied in sterile packs
  • Three sizes: 4,7 mm Int. Ø×8 mm H, 6,4 mm Int. Ø×8 mm H and 9,5 mm Int. Ø×11 mm H
To isolate a clone, a thin layer of grease is applied to the bottom edge of the cylinder and then inverted over the clone of choice. After adding a small volume ofMore Product Information