Supelco® TraceCERT® Organic Reference Standard, Tricyclazole
Supplier: Merck
39116-10MG
39116-10MGEA 87.4 EUR
39116-10MG
Supelco® TraceCERT® Organic Reference Standard, Tricyclazole
Standards Organic Standards
The tricyclazole CRM may also find the following uses: Determination of tricyclazole content in paddy rice using sliver colloid as a substrate and pyridine as an internal standard by surface-enhanced Raman spectroscopy (SERS). To detect residues of tricyclazole and thiram in pear fruit extracts using SERS. Dissipation studies of penoxsulam tricyclazole and profoxydim in rice paddy systems using HPLC-Diode Array Detection (DAD). Determination of tricyclazole residues in rice using liquid chromatography-tandem mass spectrometry (LC-MS/MS) along with the QuEChERS method of sample preparation. Degradation of tricyclazole (TC) in an aqueous solution and real pesticide industry wastewater (RPW) by electron beam (EB) radiation.
Certifications: The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Caution: P264 - P270 - P273 - P301 + P310 - P405 - P501