Cell culture flasks, Nunclon™ Sphera™
Supplier: Thermo Fisher Scientific
174951 174952
392-0379EA 351 EUR
392-0379 392-0380
Cell culture flasks, Nunclon™ Sphera™
Flasks Cell Culture Flasks
The Nunclon™ Sphera™ culture surface allows cells to grow in suspension with virtually no cell attachment. The surface supports many different cell types and their ability to generate spheroids. Studies show that over time the spheroids grow in volume, indicating that the new surface enables consistent, repeatable cell growth.
  • Optimises spheroid formation
  • Supports the consistent formation of cancer spheroids that simulate the 3D structures of tumour growth
  • Promotes formation of embryoid bodies (EB)
The Nunclon™ Sphera™ surface minimises variabilityMore Product Information