Bismuth (III) phosphate ≥99%

Supplier: Thermo Fisher Scientific

39380.18 39380.30
39380.18EA 108 EUR
39380.18 39380.30
Bismuth (III) phosphate ≥99%
Bismuth (III) phosphate

Purity: 99% CAS No.: 10049-01-1 Molecular Formula: BiPO4 Formula Weight: 303.95 Physical Form: Powder Density: 6.32 Odour: Odourless Melting Point: dec. Solubility: Insoluble in water and alkali. Soluble in hydrochloric acid MDL No.: MFCD00049721

Formula: BiO₄P
Melting Pt: Dec.
Density: 6,32
Storage Temperature: Ambient
MDL Number: MFCD00049721
CAS Number: 10049-01-1
EINECS: 233-161-2
Merck Index: 14,1275

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR