Indium(I) bromide, ultra dry ≥99.998% (metals basis)

Supplier: Thermo Fisher Scientific
Danger


ampouled under argon

39388.14
39388.14EA 1000 EUR
39388.14
Indium(I) bromide, ultra dry ≥99.998% (metals basis)
Indium (I) bromide

Purity: 99.998% CAS No.: 14280-53-6 Molecular Formula: InBr Formula Weight: 194.73 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 4.96 Melting Point: 220° Boiling Point: 662° UN No.: 3260 MDL No.: MFCD00064630

Formula: InBr
Boiling Pt: 662 °C
Melting Pt: 220 °C
Density: 4,96
Storage Temperature: Ambient
MDL Number: MFCD00064630
CAS Number: 14280-53-6
EINECS: 238-172-6
UN: 3260
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR