Tetrakis(acetonitrile)palladium(II) tetrafluoroborate (23,5% Pd)

Supplier: Thermo Fisher Scientific
Warning

39446.01 39446.03
39446.01EA 74.6 EUR
39446.01 39446.03
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate (23,5% Pd)
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate

CAS No.: 21797-13-7 Molecular Formula: [Pd(CH3CN)4](BF4)2 Formula Weight: 444.22 Physical Form: Powder Sensitivity: Air Sensitive Melting Point: 230° dec. Solubility: Soluble in water UN No.: UN2811 MDL No.: MFCD00043297

Formula: C₈H₁₂B₂F₈N₄Pd
MW: 444.239 g/mol
Melting Pt: 230 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00043297
CAS Number: 21797-13-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR