Periodic acid solution, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY
Danger

Synonyms: ortho-Periodic acid, Orthoperiodic acid

3951-100ML
3951-100MLEA 109 EUR
3951-100ML
Periodic acid solution, Sigma-Aldrich®
Periodic acid

For use in procedure no. 395 for Periodic Acid-Schiff (PAS) and Silver Stain Procedure No. HT100.

Formula: H₅IO₆
MW: 227.94 g/mol
Melting Pt: 122
Storage Temperature: Refrigerator
MDL Number: MFCD00011344
CAS Number: 10450-60-9
ADR: 8 II
Merck Index: 13,07248

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR