Cobalt 99,95 Metals basis, foil, Thickness: 0.025 mm (0.001 in)

Supplier: Thermo Fisher Scientific
Danger

40183.FF 40183.FI
40183.FFEA 237 EUR
40183.FF 40183.FI
Cobalt 99,95 Metals basis, foil, Thickness: 0.025 mm (0.001 in)
Cobalt

Purity: 99.95% CAS No.: 7440-48-4 Physical Form: ≈0.14g/25x25mm Odour: Odourless MDL No.: MFCD00010935

Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR