Aluminium oxide, single crystal

Supplier: Thermo Fisher Scientific

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

40261.KG 40261.KJ
40261.KGEA 72.4 EUR
40261.KG 40261.KJ
Aluminium oxide, single crystal
Aluminium oxide

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: 12.7mm dia x 1.0mm thick, random orientation MDL No.: MFCD00003424

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR