Magnesium ≥99.9% (metals basis), foil, hard, Thickness: 0.125 mm (0.005 in)

Supplier: Thermo Fisher Scientific
Danger

40602.FF 40602.FI
40602.FFEA 285 EUR
40602.FF 40602.FI
Magnesium ≥99.9% (metals basis), foil, hard, Thickness: 0.125 mm (0.005 in)
Magnesium

Purity: 99.9% CAS No.: 7439-95-4 Physical Form: ≈0.14g/25x25mm, Temper: as rolled Odour: Odourless UN No.: 1869 MDL No.: MFCD00085308

Formula: Mg
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1869
ADR: 4.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR