Strontium-2-ethylhexanoate 40% (8 - 12% Sr) in 2-ethylhexanoic acid

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 2-Ethylhexanoic acid strontium salt

40719.06 40719.14 40719.22
40719.06EA 0 EUR
40719.06 40719.14 40719.22
Strontium-2-ethylhexanoate 40% (8 - 12% Sr) in 2-ethylhexanoic acid
Strontium 2-ethylhexanoate

CAS No.: 2457-02-5 Molecular Formula: C16H30O4Sr Formula Weight: 374.04 Physical Form: Liquid Odour: Mild MDL No.: MFCD00070438

Formula: C₁₆H₃₀O₄Sr
MW: 374.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070438
CAS Number: 2457-02-5
EINECS: 219-536-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR