Ionomycin, Millipore®

Supplier: Merck Millipore Calbiochem

407950-1 407950-10 407950-5
407950-1EA 460 EUR
407950-1 407950-10 407950-5
Ionomycin, Millipore®
Ionomycin
Formula: C₄₁H₇₂O₉
MW: 709.02 g/mol
MDL Number: MFCD06798385
CAS Number: 56092-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR