Germanium-n-butoxide ≥97%

Supplier: Thermo Fisher Scientific
Warning

40816.03 40816.06 40816.14
40816.03EA 55.8 EUR
40816.03 40816.06 40816.14
Germanium-n-butoxide ≥97%
Germanium-n-butoxide

Purity: 97+% CAS No.: 25063-27-8 Molecular Formula: C16H36GeO4 Formula Weight: 365.05 Flash Point: 112°(234°F) Physical Form: Liquid Density: 1.017 Refractive Index: 1.4255 Sensitivity: Moisture Sensitive Boiling Point: 143°/8mm MDL No.: MFCD00015347


Applications: Oxide films

Formula: C₁₆H₃₆GeO₄
Boiling Pt: 143 °C (8 mmHg)
Density: 1,017
Flash Pt: 112 °C (234 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015347
CAS Number: 25063-27-8
EINECS: 246-595-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR