Cobalt 99,95 Metals basis, sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)

Supplier: Thermo Fisher Scientific
Danger

40837.KS
40837.KSEA 0 EUR
40837.KS
Cobalt 99,95 Metals basis, sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Cobalt

Purity: 99.95% CAS No.: 7440-48-4 Odour: Odourless MDL No.: MFCD00010935

Formula: Co
MW: 58.93 g/mol
Boiling Pt: 2870
Melting Pt: 1495
Density: 8.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010935
CAS Number: 7440-48-4
EINECS: 231-158-0
Merck Index: 12,02488

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR