Supelco® Organic Reference Standard, β-Alanine

Supplier: Merck

41144-100MG
41144-100MGEA 52.8 EUR
41144-100MG
Supelco® Organic Reference Standard, β-Alanine
Standards Organic Standards

β-Alanine flux plays a cytoprotective role by supporting the osmotic stability of marine organisms, preimplantation mouse embryos and mammalian cells exposed to hypoxic stress.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d092050143c07b525fd1579c31a5003e7184d853&vid=e9f3e624808031db86d489004ea3d162cb6d6ba4&prid=web&clid=SAPDAM

Order Now

SPECIFICATIONS

Synonym(s) β-Ala; 3-Aminopropionic acid
Environmentally Preferable
M-Clarity quality level MQ100
CAS No. 107-95-9
MDL No. MFCD00008200
Formula NH₂CH₂CH₂COOH
Molecular weight 89,09
Melting point 202 °C (dec.) (lit.)
Flash point Not applicable
Shipping conditions STD
Storage conditions 2...30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR