Vortex mixer, VORTEX Genius 3
Supplier: IKA
3340000 3340002 3340003
412-0098EA 448 EUR
412-0098 412-0115 412-0116
Vortex mixer, VORTEX Genius 3
Shakers Vortex Shakers
Vortex mixer with three interchangeable attachments and various inserts for different applications. Microcentrifuge tubes, microtitre plates, even round bottomed or 250 ml Erlenmeyer flasks can be attached by means of the specially designed strap. Attachments click securely onto the unit in any position. The unit is stable at high speeds and sturdy with coated, zinc die-casting.
  • Compact design
  • Wide speed range, infinitely adjustable
  • Touch or continuous mode
  • Can be used at 5 to 40 °C (80% relative humidity) in an incubator or cold room
IP protectionMore Product Information