Microfine grinder, MF 10 basic
Supplier: IKA
2836000 2870900 2871000 2836002 2836003
412-1130EA 4890 EUR
412-1130 412-1131 412-1132 412-1128 412-1129
Microfine grinder, MF 10 basic
Mills
Continuously operating universal grinder with powerful drive. When using the grinding heads, the processed material passes through an interchangeable stainless steel sieve. The ground material can be collected using a vessel with NS 29 standard ground joint.
  • Easy to clean stainless steel working surface
  • Choice of two different heads that can be used with the drive
  • Heads are easily interchanged
IP protection class according to DIN EN 60529: IP 22
Certifications: IP protection class according to DIN EN 60529: IP 22
Ordering information: More Product Information