Silver 99,99 Metals basis, sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)

Supplier: Thermo Fisher Scientific

41267.KS
41267.KSEA 0 EUR
41267.KS
Silver 99,99 Metals basis, sputtering target, Ø 76.2 mm (3.0 in), Thickness: 6.35 mm (0.250 in)
Silver

Purity: 99.99% CAS No.: 7440-22-4 Physical Form: Rolled metal disc ~305grams Odour: Characteristic MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR