Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt 30% (w/v) in aqueous solution

Supplier: Thermo Fisher Scientific

41518.K2 41518.AD
41518.K2EA 0 EUR
41518.K2 41518.AD
Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt 30% (w/v) in aqueous solution
Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt

Molecular Formula: C26H60N6O8 Formula Weight: 276.44 Physical Form: Liquid MDL No.: MFCD01632148


Applications: Chelating agent, polymerization catalyst, detergent

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR