Titanium ≥99.9% (metals basis), spherical powder -150 mesh

Supplier: Thermo Fisher Scientific
Danger

41545.14 41545.22 41545.36
41545.14EA 59.4 EUR
41545.14 41545.22 41545.36
Titanium ≥99.9% (metals basis), spherical powder -150 mesh
Titanium

Purity: 99.9% CAS No.: 7440-32-6 Odour: Odourless UN No.: 2546 MDL No.: MFCD00011264

Formula: Ti
Boiling Pt: 3277 °C (1013 hPa)
Melting Pt: 1677 °C
Density: 4,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
UN: 2546
ADR: 4.2,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR