Silver 99,9 Metals basis, powder_spherical APS 1.3-3.2 µm

Supplier: Thermo Fisher Scientific

41599.09 41599.18 41599.30
41599.09EA 79.2 EUR
41599.09 41599.18 41599.30
Silver 99,9 Metals basis, powder_spherical APS 1.3-3.2 µm
Silver

Purity: 99.9% CAS No.: 7440-22-4 Physical Form: S.A. 0.3-0.7m2/g Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397

Formula: Ag
MW: 107.87 g/mol
Boiling Pt: 2212
Melting Pt: 962
Density: 10.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003397
CAS Number: 7440-22-4
EINECS: 231-131-3
UN: 3077
ADR: 9 III
Merck Index: 12,08647

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR