(±)-2-Butanol, anhydrous 99%

Supplier: Thermo Fisher Scientific
Warning


Packaged under argon in resealable ChemSeal™ bottles.

41871.AK 41871.K2
41871.AKEA 40.6 EUR
41871.AK 41871.K2
(±)-2-Butanol, anhydrous 99%
(±)-2-Butanol

Purity: 99% CAS No.: 78-92-2 Molecular Formula: C4H10O Formula Weight: 74.12 Flash Point: 26°(79°F) Physical Form: Liquid Density: 0.808 Refractive Index: 1.3970 Sensitivity: Hygroscopic Appearance: Colourless Odour: Sweet Melting Point: -115° Boiling Point: 98-101° Beilstein Reference: 1718765 Merck Reference: 14,1541 UN No.: 1120 MDL No.: MFCD00004569

Formula: H₃CCH₂CH(OH)CH₃
Boiling Pt: 98…101 °C
Melting Pt: –115 °C
Density: 0,808
Flash Pt: 26 °C (79 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3,II
Merck Index: 14,1541
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1dea3607167631fbfcf7751c20b71f345ad24c19&vid=010e04e8f508fd3d501873d20c2349817834b6e8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR