di-Sodium succinate hexahydrate ≥99%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Sodium succinate hexahydrate, Succinic acid disodium hexahydrate, 1,4-Butanedioic acid disodium salt hexahydrate, Sodium Succinate, Hexahydrate, Sodium succinate dibasic hexahydrate, Disodium succinate hexahydrate, Succinic acid disodium salt hexahydrate

41983.30 41983.A1 41983.A7
41983.30EA 66.5 EUR
41983.30 41983.A1 41983.A7
di-Sodium succinate hexahydrate ≥99%
di-Sodium succinate hexahydrate

Purity: 99% CAS No.: 6106-21-4 Molecular Formula: C4H4Na2O4.6H2O Formula Weight: 270.14 (162.05anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: White Melting Point: 120° -6H2O Beilstein Reference: 3921044 Merck Reference: 14,8679 MDL No.: MFCD00149117

Formula: NaOOCCH₂CH₂COONa·6H₂O
Melting Pt: 120 °C (-6H₂O)
Density: 1,529 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149117
CAS Number: 6106-21-4
EINECS: 205-778-7
Merck Index: 14,8679

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR