IPTG (isopropyl-β-D-thiogalactopyranoside), Millipore®

Supplier: Merck Millipore Calbiochem

420322-1 420322-10 420322-25 420322-5
420322-1EA 236 EUR
420322-1 420322-10 420322-25 420322-5
IPTG (isopropyl-β-D-thiogalactopyranoside), Millipore®
IPTG (Isopropyl-β-D-thiogalactopyranoside)
Formula: C₉H₁₈O₅S
Boiling Pt: 402 °C (1013 hPa)
Melting Pt: 109,5…110.5 °C
Storage Temperature: Freezer
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR