L-Lysine, anhydrous 98,5-102,0, bioreagent for biopharmaceutical production

Supplier: VWR Chemicals

Synonyms: (S)-2,6-Diaminohexanoic acid, L(+)-Lysine, (S)-(+)-2,6-Diaminohexanoic acid, (S)-(+)-Lysine, (S)-(+)-2,6-Diaminocaproic acid, H-Lys-OH

42307-1KGEA 0 EUR
42307-1KG
L-Lysine, anhydrous 98,5-102,0, bioreagent for biopharmaceutical production
L-Lysine
Formula: C₆H₁₄N₂O₂
MW: 146.19 g/mol
Boiling Pt: 290
Melting Pt: 196
MDL Number: MFCD00064433
CAS Number: 56-87-1
EINECS: 200-294-2
Merck Index: 13,05656

Order Now

Specification Test Results

Assay (on dried substance) 98.5 - 102.0 %
Appearance Crystalline powder
Colour White to off white
Cell culture toxicity Passes test
DNases Not detected
Identification by IR Conforms to structure
Proteases Not detected
RNases Not detected
Solubility (10 %) Clear.complete.yellowish
Spec. opt. rot.(25°C; 8 %; HCl 6N;dried) 25.5 - 26.7 °
Heavy metals (as Pb) Max. 15 ppm
Ignition residue Max. 0.1 %
Water Max. 2.0 %
SO₄ (Sulphate) Max. 0.03 %
Fe (Iron) Max. 30 ppm
Bioburden Max. 100 CFU/g
Endotoxin (1 %) (EU/mg) Max. 0.1 EU/mg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR