Glassy carbon, powder 2000-3150 µm Type2

Supplier: Thermo Fisher Scientific
Warning

42515.A1
42515.A1EA 0 EUR
42515.A1
Glassy carbon, powder 2000-3150 µm Type2
Glassy carbon
Formula: C
MW: 12.01 g/mol
Melting Pt: 3652
Density: 1.95 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00133992
CAS Number: 7440-44-0
EINECS: 231-153-3
UN: 1325
ADR: 4.1 III
Merck Index: 13,01818

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR