Dimethyl carbonate, extra dry 99+%, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

428730010 428731000
428730010.EA 184 EUR
428730010. 428731000.
Dimethyl carbonate, extra dry 99+%, AcroSeal®
Dimethyl carbonate
Formula: C₃H₆O₃
Boiling Pt: 90 °C (1013 hPa)
Melting Pt: 2…4 °C
Density: 1,069 g/cm³
Flash Pt: 18 °C
Storage Temperature: Ambient
MDL Number: MFCD00008420
CAS Number: 616-38-6
UN: 1161
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %
Color scale ≤30 APHA

SPECIFICATIONS

Environmentally Preferable Safer Human and Environmental Health
Environmental certification Chem21 Solvent Selection Guide recommended by default and after discussion

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR