Multi-element plasma standard solution 1, Specpure®

Supplier: Thermo Fisher Scientific

42881.AEEA 379 EUR
42881.AE
Multi-element plasma standard solution 1, Specpure®
Standards Cationic Standards Multi-Element Standards

UN No.: UN3264
MDL No.: MFCD00212337

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR