Zinc acetate dihydrate 99.0-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Warning

4296-01EA 100.07 EUR
4296-01 4296-07 4296-R
Zinc acetate dihydrate 99.0-101.0%, crystals, BAKER ANALYZED® ACS, J.T.Baker®
Zinc acetate dihydrate

Crystals. Lot analysis on label.


  • Lot analysis on label

Formula: (H₃CCOO)₂Zn·2H₂O
Melting Pt: 237 °C
Density: 1,74 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00066961
CAS Number: 5970-45-6
EINECS: 209-170-2
UN: 3077
ADR: 9,III
Merck Index: 12,10256

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements 99.0 - 101.0%
For Laboratory,Research,or Manufacturing Use ≤0.005%
ACS - Assay ((CH₃COO)₂Zn·2H₂O) 99.0 - 101.0 %
ACS - Insoluble Matter ≤0.005 %
ACS - pH of 5% Solution at 25°C 6.0 - 7.0
ACS - Sulfate (SO₄) ≤0.002 %
ACS - Calcium (Ca) ≤0.005 %
ACS - Lead (Pb) ≤0.002 %
ACS - Magnesium (Mg) ≤0.005 %
ACS - Potassium (K) ≤0.01 %
ACS - Sodium (Na) ≤0.05 %
Chloride (Cl) ≤5 ppm
Trace Impurities - Iron (Fe) ≤5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR