Borane dimethyl sulphide complex 2 M in toluene

Supplier: Thermo Fisher Scientific
Danger


Packaged under argon in resealable ChemSeal™ bottles.

42963.AE 42963.AP
42963.AEEA 155 EUR
42963.AE 42963.AP
Borane dimethyl sulphide complex 2 M in toluene
Borane dimethyl sulphide complex

CAS No.: 13292-87-0 Molecular Formula: C2H9BS Formula Weight: 75.97 Flash Point: 7°(45°F) Physical Form: Liquid Density: 0.856 Appearance: Colourless Beilstein Reference: 3663489 UN No.: 3399 MDL No.: MFCD00013189

Formula: C₂H₉BS
Density: 0,856
Flash Pt: 7 °C (45 °F)
Storage Temperature: Ambient
MDL Number: MFCD00013189
CAS Number: 13292-87-0
EINECS: 236-313-6
UN: 3399
ADR: 4.3,I

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR