Cesium acetate ≥99% (metals basis)

Supplier: Thermo Fisher Scientific

43095.30 43095.18
43095.30EA 288 EUR
43095.30 43095.18
Cesium acetate ≥99% (metals basis)
Cesium acetate

Purity: 99% CAS No.: 3396-11-0 Molecular Formula: CsOOCCH3 Formula Weight: 191.95 Physical Form: Crystalline Powder Odour: Weak MDL No.: MFCD00013056

Formula: H₃CCOOCs
Melting Pt: 194 °C
Storage Temperature: Ambient
MDL Number: MFCD00013056
CAS Number: 3396-11-0
EINECS: 222-248-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR