Chromium(III) nitrate nonahydrate ≥99.99% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Chromium trinitrate nonahydrate, Chromic nitrate nonahydrate, Chromium nitrate nonahydrate

43182.22 43182.14 43182.06
43182.22EA 779 EUR
43182.22 43182.14 43182.06
Chromium(III) nitrate nonahydrate ≥99.99% (metals basis)
Chromium (III) nitrate nonahydrate

Purity: 99.99% min CAS No.: 7789-02-8 Molecular Formula: Cr(NO3)3.9H2O Formula Weight: 400.14 (238.01anhy) Physical Form: Crystalline Appearance: Purple Odour: Odourless Melting Point: 60° Merck Reference: 14,2226 UN No.: 2720 MDL No.: MFCD00149659

Formula: CrN₃O₉·9H₂O
Melting Pt: 60 °C
Density: 1,807 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149659
CAS Number: 7789-02-8
EINECS: 236-921-1
UN: 2720
ADR: 5.1,III
Merck Index: 14,2226

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR