Probes and accessories for Vibra-Cell™ ultrasonic processors
Supplier: SONICS MATERIALS
630-0310 630-0409 630-0418 630-0731 630-0444 830-00004 630-0207 630-0208 630-0210 630-0209 630-0421 830-00118 830-00116 630-0423 630-0435 630-0419 830-00060 630-0220 630-0217 630-0218 630-0306 630-0420 630-0219 630-0559 630-0579 630-0422 630-0580 630-0608 630-0406 630-0651 630-0407 830-00130
SONI630-0310EA 1040 EUR
SONI630-0310 432-0111 432-0112 432-0117 432-0119 432-0128 432-0102 432-0103 432-0105 432-0104 432-0115 432-0131 432-0130 432-0116 432-0118 432-0113 432-0129 432-0107 SONI630-0217 SONI630-0218 SONI630-0306 432-0114 432-0106 432-0123 432-0125 432-0127 SONI630-0580 SONI630-0608 432-0267 432-0269 SONI630-0407 SONI830-00130
Probes and accessories for Vibra-Cell™ ultrasonic processors
Homogenisers Ultrasonic Cell Disruptors
A wide range of probes for a variety of applications is available, including stepped and tapered microtips and also probes for processing in microplates.
  • Multi-element probes for deep well microplates
  • Cuphorns for non invasive sonication of samples
  • Continouous flow cells for dispersing and homogenising large volumes of low viscosity samples where extended exposure to ultrasonics is not desirable
  • Footswitch for hands-free operation
  • Sound abating enclosures - reduce noise and act as a containment facility
Probes for processing in microplates:
The multi-More Product Information