Platinum ≥99.95% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)

Supplier: Thermo Fisher Scientific

Synonyms: Platinium

43276.BQ 43276.BY 43276.BU
43276.BQEA 239 EUR
43276.BQ 43276.BY 43276.BU
Platinum ≥99.95% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)
Platinum

Purity: 99.95% CAS No.: 7440-06-4 Physical Form: ≈16.8g/m, Temper: soft Odour: Odourless MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Density: 21.447 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
Merck Index: 12,07689

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR