Grinding pieces, zirconium oxide, yttria stabilised

Supplier: Thermo Fisher Scientific
43286.A1 43285.A1 43286.30 43285.30
43286.A1EA 542 EUR
43286.A1 43285.A1 43286.30 43285.30
Grinding pieces, zirconium oxide, yttria stabilised
Beads Ceramic Beads
The density of the zirconium oxide beads is 6.02 g/cm³

  • MDL No.: MFCD00145546
  • Yttrium oxide (mole) 3%
  • Hardness 12
  • Total impuroty level < 0.1

Storage at ambient temperatures.
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR