Gold trichloride ≥99.99% (metals basis) Au ≥ 64.4%, Premion®

Supplier: Thermo Fisher Scientific
Danger

43360.03 43360.06
43360.03EA 326 EUR
43360.03 43360.06
Gold trichloride ≥99.99% (metals basis) Au ≥ 64.4%, Premion®
Gold trichloride

Purity: 99.99% CAS No.: 13453-07-1 Molecular Formula: AuCl3 Formula Weight: 303.33 Physical Form: Crystalline Density: 3.90 Appearance: Red Odour: Odourless Melting Point: 180(subl) Merck Reference: 14,4521 UN No.: 3260 MDL No.: MFCD00014172

Formula: AuCl₃
Melting Pt: 180 °C (subl.)
Density: 3,90
Storage Temperature: Ambient
MDL Number: MFCD00014172
CAS Number: 13453-07-1
EINECS: 236-623-1
UN: 3260
ADR: 8,III
Merck Index: 14,4521

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR