Lithium wolframate ≥99%

Supplier: Thermo Fisher Scientific

43366.30 43366.18
43366.30EA 496 EUR
43366.30 43366.18
Lithium wolframate ≥99%
Lithium wolframate

Purity: 99% min CAS No.: 13568-45-1 Molecular Formula: Li2WO4 Formula Weight: 261.73 Physical Form: Powder Density: 3.71 Melting Point: 742° MDL No.: MFCD00016185

Formula: Li₂O₄W
Melting Pt: 742 °C
Density: 3,71
Storage Temperature: Ambient
MDL Number: MFCD00016185
CAS Number: 13568-45-1
EINECS: 236-978-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR