5-Methylcytosine ≥97%

Supplier: Thermo Fisher Scientific

Synonyms: 4-Amino-5-methylpyrimidin-2(1H)-one

43492.MCEA 142 EUR
43492.MC 43492.ME
5-Methylcytosine ≥97%
5-Methylcytosine

Purity: 97% CAS No.: 554-01-8 Molecular Formula: C5H7N3O Formula Weight: 125.13 Physical Form: Crystalline MDL No.: MFCD00233537

Formula: C₅H₇N₃O
Storage Temperature: Ambient
MDL Number: MFCD00233537
CAS Number: 554-01-8
EINECS: 209-058-3
Merck Index: 14,6050

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR