STYRENE, dry, latex microsphere 1 micron

Supplier: Thermo Fisher Scientific

43711.MEEA 770 EUR
43711.ME
STYRENE, dry, latex microsphere 1 micron
Poly(styrene)

CAS No.: 9003-53-6 Molecular Formula: [-CH2CH(C6H5)-]n Odour: Odourless MDL No.: MFCD00084450

Formula: C₈H₈
MW: 125000 - 250000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00084450
CAS Number: 9003-53-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR