1-Propanol ≥99.5% ACS

Supplier: Thermo Fisher Scientific
Danger

43848.K2 43848.K7 43848.AP 43848.M6
43848.K2EA 89.5 EUR
43848.K2 43848.K7 43848.AP 43848.M6
1-Propanol ≥99.5% ACS
1-Propanol

Purity: 99.5+% CAS No.: 71-23-8 Molecular Formula: CH3CH2CH2OH Formula Weight: 60.10 Flash Point: 15°(59°F) Physical Form: Liquid Density: 0.804 Refractive Index: 1.3850 Odour: Alcohol-like Melting Point: -127° Boiling Point: 97° Beilstein Reference: 1098242 Merck Reference: 14,7842 UN No.: 1274 MDL No.: MFCD00002941

Formula: H₃CCH₂CH₂OH
Boiling Pt: 97 °C
Melting Pt: –127 °C
Density: 0,804
Flash Pt: 15 °C (59 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002941
CAS Number: 71-23-8
EINECS: 200-746-9
UN: 1274
ADR: 3,II
Merck Index: 14,7842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab459f809f7797b46eb6d275aa3b1e6e94dc70d1&vid=deb28b985024604344065eea93ff6c6a9dd5a431&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR