Potassium tetrachloroplatinate(II) ≥99.99% (metals basis) Pt ≥ 46.0%, Premion®

Supplier: Thermo Fisher Scientific
Danger

43946.03 43946.06 43946.14
43946.03EA 201 EUR
43946.03 43946.06 43946.14
Potassium tetrachloroplatinate(II) ≥99.99% (metals basis) Pt ≥ 46.0%, Premion®
Potassium tetrachloroplatinate (II)

Purity: 99.99% CAS No.: 10025-99-7 Molecular Formula: Cl4K2Pt Formula Weight: 415.11 Physical Form: Crystalline Density: 3.38 Appearance: Red-brown Odour: Odourless Melting Point: dec. Merck Reference: 14,7682 UN No.: 3288 MDL No.: MFCD00011378

Formula: Cl₄K₂Pt
Boiling Pt: 35…40 °C (20 mmHg)
Melting Pt: Dec.
Density: 3,38
Storage Temperature: Ambient
MDL Number: MFCD00011378
CAS Number: 10025-99-7
EINECS: 233-050-9
UN: 3288
ADR: 6.1,III
Merck Index: 14,7682

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR