Lithium acetylide ethylenediamine complex 25% w/w slurry in toluene, AcroSeal®

Supplier: Thermo Fisher Scientific
Danger

439881000
439881000.EA 374 EUR
439881000.
Lithium acetylide ethylenediamine complex 25% w/w slurry in toluene, AcroSeal®
Lithium acetylide ethylenediamine complex
Formula: C₄H₉LiN₂
MW: 92.07 g/mol
Flash Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00013183
CAS Number: 6867-30-7
UN: 3399
ADR: 4.3 I

Order Now

Specification Test Results

Appearance Tan to orange-brown Slurry
Titration with HClO4 23.5 to 30.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR