Chromium(III) potassium sulphate dodecahydrate ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Warning

44083.14 44083.22
44083.14EA 91.8 EUR
44083.14 44083.22
Chromium(III) potassium sulphate dodecahydrate ≥99.9% (metals basis)
Chromium (III) potassium sulphate dodecahydrate

Purity: 99.9% CAS No.: 7788-99-0 Molecular Formula: CrKO8S2.12H2O Formula Weight: 499.39 (283.21anhy) Physical Form: Crystalline Density: 1.83 Appearance: Dark purple Odour: Odourless Melting Point: 89° Merck Reference: 14,2229 MDL No.: MFCD00149917

Formula: CrK(SO₄)₂·12H₂O
Melting Pt: 89 °C
Density: 1,83
Storage Temperature: Ambient
MDL Number: MFCD00149917
CAS Number: 7788-99-0
EINECS: 233-401-6
Merck Index: 14,2229

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR